FlashPure ID Silica and C18 Flash Cartridges, Büchi
Supplier: Buchi
145146133 145152104 145152105 145146130 145146131 145165730 145171532 145146133 145152104 145152105 145146130 145146131
75794-620EA 735.08 CAD
75794-620 75794-622 75794-624 75794-626 75794-628 75794-630 75794-632 75794-634 75794-688 75794-690 75794-692 75794-744 75794-746 75794-748 75794-750 75794-752 75794-754 75794-756 75794-758 75794-760 75794-762 75794-764 75794-766 75794-768 75794-786 75794-788 CA75794-620 CA75794-626 CA75794-628 CA75794-630 CA75794-632 CA75794-688 CA75794-690 CA75794-692
FlashPure ID Silica and C18 Flash Cartridges, Büchi
Chromatography Columns
Standard Silica Flash Cartridges are used for optimum chromatographic performance.
  • High resolution of separation due to high quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • The RFID tag tracks cartridge usage and history
Superior Resolution with ultrapure 40 μm or silicaStandard and High Performance cartridges available ∙ Silica and multiple bonded phasesMore Product Information