Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Supplier: Simport Scientific
T312-8 T312-10 T312-4 T312-7 T312-11 T312-5 T312-3 T312-9 T312-2 T312-1 T312-14 T312-13 T312-5 T312-7 T312-8 T312-9
CA73960-401CS 29.83 CAD
CA73960-401 CA73960-407 CA73960-408 CA73960-409 CA73960-413 CA73960-406 CA73960-410 CA73960-411 CA73960-412 CA73960-414 CA73960-415 CA73960-417 490010-380 490010-382 490010-384 490010-386
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Cap Inserts
Color coded inserts fit precisely into the cap of the Cryovial® tubes for color identification.
Made of polypropylene
Ordering information: T312-7 contains 100 pieces each of white, blue, red, green and yellow inserts packaged in individual bags.