Histosette® II Tissue Processing/Embedding Cassettes with Integrated Lids, Simport
Supplier: Simport Scientific
M492-16 M492-10 M492-11 M492-2 M492-3 M492-4 M492-5 M492-7 M492-8 M492-9 M492-18
CA73960-512CS 509.98 CAD
CA73960-512 CA73960-514 CA73960-054 CA73960-056 CA73960-060 CA73960-062 CA73960-064 CA73960-066 CA73960-068 CA73960-070 CA73960-072 CA11031-920 10040-366
Histosette® II Tissue Processing/Embedding Cassettes with Integrated Lids, Simport
Embedding Cassettes Tissue Processing Cassettes
These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage
Molded from a special high-density polymer, they can keep specimens safely submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lids can be opened and closed as often as necessary and will always relock securely without danger of specimen loss. The unique front hinge design allows for single-hand cassette manipulation. The anterior writingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable