Nunc® InVitro Roller Bottles, PETG, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
1060-20 1760-20 1060-05 2160-05 2160-20 1860-22 4260-22 1060-85 1860-22
CA44200-012CS 537.69 CAD
CA44200-012 CA62404-388 CA73520-410 CA73520-418 CA73520-420 CA73520-732 CA73520-734 CA73521-060 73520-732
Nunc® InVitro Roller Bottles, PETG, Sterile, Thermo Scientific
Bottles Roller Bottles
Tough, flexible PETG roller bottles offer a safer, more convenient alternative to conventional polystyrene roller bottles.
  • Bottles include a 48 mm standard cap, except for the bottles with a vented cap
  • Printed, easy-to-read graduations aid in the measurement of medium fills
Standard surface bottles offer 1050 cm² of surface area, providing more capacity than 850 cm² bottles for adherent or suspension cultures. They can also be used as freezing containers in temperatures as low as –40 °C (–40 °F).
Expanded surface roller bottles have specially-More Product Information