Multitrans™ Collection and Transportation Systems
Supplier: Starplex
S160-FL-COM S160-0 S160-FL S160-NASO S160-100 S160-NASO-1 S160-0
CA11006-070CS 566.82 CAD
CA11006-070 CA66410-062 CA73270-054 CA73270-056 CA73270-008 CA89128-688 66410-062 CA76636-550
Multitrans™ Collection and Transportation Systems
Swabs Transport Swabs
The Multitrans™ System is used for the collection and transportation of viruses, Chlamydia, and mycoplasmas obtained from clinical specimens within a 72 to 96 hour period.
The Multitrans medium and glass beads are held within a durable, polypropylene vial and capped with a skirted, swab retaining cap. The swab is scored so that it is easily snapped off into the vial; after capping and reopening, the swab is neatly inserted into the cap for easy, contamination free handling. The media vial and sterile wrapped swab are packaged in a plastic peel pack.
The generous amount of mediumMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable