Histoplex™ Histology Containers, Starplex®
Supplier: Starplex
B402-VO H2502-C H5002-C H10002-C H20002 H50002 H80002 H200002 H202-FL H402-FL H602-FL PH10002 PH5002 H902-FL H95002
CA66410-049CS 254.43 CAD
CA66410-049 CA73240-048 CA73240-052 CA73240-058 CA73240-062 CA73240-064 CA73240-066 CA73240-068 CA73240-090 CA73240-096 CA73240-100 CA95038-618 CA95038-620 CA73240-040L 10799-382
Histoplex™ Histology Containers, Starplex®
Sample Containers General Purpose Containers
These nonsterile containers are carefully engineered for the storage of histology specimens
The 20mL, 40mL, 60mL, and 90mL containers are clear polypropylene and have graduations for easy fill line viewing. They feature a yellow, molded O-Ring screw-on cap, which forms a seal that eliminates most formalin leakage and evaporation. They also feature reinforced top and bottom, and a full circle cap thread to eliminate cross-threading. Containers are straight-sided to save space in storage areas. The 250mL, 500mL, and 1000mL containers are the same except they feature a ribbed cap that isMore Product Information