Disodium hydrogen phosphate, anhydrous ≥99.0% ACS

Supplier: VWR

84486-300
CA71008-786EA 213.9 CAD
CA71008-786
Disodium hydrogen phosphate, anhydrous ≥99.0% ACS
di-Sodium hydrogen phosphate
Formula: Na₂HPO₄
Melting Pt: 250 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Order Now

Specification Test Results

Asssay (Na2HPO4) 99.0% min.
pH of a 5% Solution at 25°C 8.7 - 9.3
Insoluble Matter 0.01% max.
Loss on drying at 105°C 0.2% max.
Chloride (Cl) 0.002% max.
Sulfate (SO4) 0.005% max.
Heavy metals (as Pb) 0.001% max.
Iron (Fe) 0.002% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR