N-Methyl-2-pyrrolidone (NMP), Accusolv™, distilled VWR®

Supplier: Anachemia
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

60070-540
CA71007-814EA 1813.92 CAD
CA71007-814
N-Methyl-2-pyrrolidone (NMP), Accusolv™, distilled VWR®
N-Methyl-2-pyrrolidone (NMP)

Suitable for Chromatography and Spectrophotometry.

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

UV absorbance (1 cm cell vs H2O) 285 nm 1.000 max.
UV absorbance (1 cm cell vs H2O) 300 nm 0.500 max.
UV absorbance (1 cm cell vs H2O) 325 nm 0.100 max.
UV absorbance (1 cm cell vs H2O) 350 nm 0.030 max.
UV absorbance (1 cm cell vs H2O) 400 nm 0.010 max.
Assay (GC) not corrected for water 99.5% min.
Water (H2O) 0.05% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR