Methanol 99,8 ACS

Supplier: VWR
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

56902-532 56902-540 56902-543 56902-700 56902-940
CA71007-734CS 4459.76 CAD
CA71007-734 CA71007-736 CA71007-738 CA71007-742 CA71007-744 CA71007-748
Methanol 99,8 ACS
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CH3OH) 99.8% min.
Substances darkened by H2SO4 Passes Test
Substances reducing KMnO4 Passes Test
Colour (APHA) 10 max.
Water (H2O) 0.1% max.
Residue after evaporation 0.001% max.
Solubility in water Passes Test
Titrable acid 0.0003 meq/g max.
Titrable base 0.0002 meq/g max.
Carbonyl compounds:
Acetone 0.001% max.
Formaldehyde (as HCHO) 0.001% max.
Acetaldehyde 0.001% max.
Appearance Clear

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR