Sodium acetate

Supplier: G-Biosciences

RC-089 RC-090
CA71003-462EA 193.33 CAD
CA71003-462 CA71003-464
Sodium acetate
Sodium acetate
Formula: H₃CCOONa
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR