Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemical
710-109-130
CA710-109-130EA 512 CAD
CA710-109-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Goat Anti-IgG IgA IgM Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR