Gelatin
Supplier: Spectrum Chemical Mfg. Corp.
GE105-12KG
CA700011-238EA 5579 CAD
CA700011-238
Gelatin
Proteins and Peptides
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Gelatin, 250 Bloom, Powder, NF is used pharmaceutical formulations as thickening agent and emulsifier. All spectrum chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.