6-O-Palmitoyl-L-ascorbic acid 95,0 NF

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: L-Ascorbic acid 6-palmitate, Palmitoyl-L-ascorbic acid, Ascorbyl Palmitate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl palmitate, L(+)-Ascorbyl palmitate

A1372-5KGBL A1372-500GM
CA700009-358EA 12001 CAD
CA700009-358 95031-886
6-O-Palmitoyl-L-ascorbic acid 95,0 NF
6-O-Palmitoyl-L-ascorbic acid

Ascorbyl Palmitate, NF is often used to promote flow of tableting powders and to prevent their sticking in tablet presses. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₂₂H₃₈O₇
MW: 414.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005377
CAS Number: 137-66-6

Order Now

Specification Test Results

Assay (Dried Basis) 95.0 - 100.5 %
Specific Rotation [A]D25 +21° to +24°
Loss On Drying max. 2.0 %
Residue On Ignition max. 0.1 %
Heavy Metals max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR