1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Avobenzone, 1-(4-(tert-Butyl)phenyl)-3-(4-methoxyphenyl)propane-1,3-dione

A2682-100GM A2682-25GM A2682-5GM
CA700004-106EA 1129 CAD
CA700004-106 CA700004-108 CA700004-110
1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione
1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione

Avobenzone is an oil soluble ingredient used in sunscreen products to absorb the full spectrum of UVA rays. Its ability to absorb ultraviolet light over a wider range of wavelengths than many organic sunscreen agents has led to its use in many commercial preparations marketed as 'broad spectrum' sunscreens. Avobenzone has an absorption maximum of 357 nm. It appears as an odorless, yellowish solid.

Formula: C₂₀H₂₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00210252
CAS Number: 70356-09-1

Order Now

Specification Test Results

Appearance Slightly Yellow-
Assay 97.0 % min.
Melting Point 81 - 86°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR