Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Supplier: Cardinal Health
90308 90408 90422 90713 92318 91616 91015 91218 92515 90610 92510 92308 90411 90810 91822 92923 92152 92322 90515 92713 91315 92309 92515 92713 92923 90411
CA11213-021PK 65.23 CAD
CA11213-021 CA11213-023 CA11213-027 CA11213-033 CA11213-236 CA70000-409 CA70000-414 CA70000-416 CA89045-093 CA11206-410 CA11206-414 CA11206-415 CA11213-025 CA11213-035 CA11213-044 CA11213-062 CA11213-230 CA11213-239 CA11213-243 CA11213-245 CA70000-418 CACV92309 11213-243 11213-245 11213-062 11213-025
Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Bags Sterilization Pouches
Peel-open pouches are designed for quick, easy presentation of sterile items.
  • Can be used in steam or EO sterilization
  • Chemical process indicators on every pouch identify the sterilization process used
Heat-seal pouches can be heat-sealed or taped shut. Self-seal pouches feature a pressure-sensitive closure that is prefolded to help ensure the integrity of the seal.
Items within the pouch are easily viewed through the blue-tinted 2 mil-thick film side. The other side is made of tough, moisture-resistant, surgical-grade paper, which provides highMore Product Information