Solvent Filter/Degasser, MilliporeSigma
Supplier: MilliporeSigma
692000-1
CA692000-1EA 5658.11 CAD
CA692000-1 CADM692001-1
Solvent Filter/Degasser, MilliporeSigma
Filters Liquid Process Filters
Used for HPLC.
Attaches directly to an empty, one-gallon solvent bottle. Designed to filter particulates that could damage expensive pumps and detectors and to remove dissolved oxygen in the solvent that could cause detector drift and instability. Filters and degasses one gallon of solvent in 10 minutes without operator attention.