O-Rings for Spray Chambers
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
6910009700 6910009800 6910013600 6910037000 6910000100 6910012100 9910093400 6910017400 6910004000 6910005200 6910010400 6910015200 6910036800 6910036900 9910026500 9910045700 9910093500 6910013200 6910013300 6910020800
CA6910009700EA 29.22 CAD
CA6910009700 CA6910009800 CA6910013600 CA6910037000 CA6910000100 CA6910012100 CA9910093400 CAAG6910017400 CAAG6910004000 CAAG6910005200 CAAG6910010400 CAAG6910015200 CAAG6910036800 CAAG6910036900 CAAG9910026500 CAAG9910045700 CAAG9910093500 AG6910013200 AG6910013300 AG6910020800
O-Rings for Spray Chambers
Seals Sealing Rings
O-rings for routine maintenance of the current Mark seven or earlier Mark VI flame atomization system.
  • Regular inspection of the O-rings in the flame atomization system is recommended
  • O-rings should be replaced if they are perished, show excessive wear or have any nicks or cuts on the surface
  • The regular black O-rings are suitable for use with aqueous solutuions, including acid digests
  • For compatibility with organic solvents, perfluoroelastomer (white) O-rings should be fitted.
  • The perfluoroelastomer O-rings are suitable for use with both aqueous and
More Product Information