Starswabs II® Specimen Collection and Transport Systems
Supplier: Starplex
SP130L SP130L-D SP130L-W SP131D SP130-W SP132-W S09 SP131-W SP130D SP132D SP132 SP130X SP131X SP130L-D
CA66410-022CS 640.68 CAD
CA66410-022 CA66410-026 CA66410-028 CA66410-032 CA66410-051 CA66410-057 CA73270-010 CA73270-021 CA66410-101 CA73270-003 CA73270-005 CA73270-052 CA66410-106L CA66410-103L CA66410-105L 66410-026
Starswabs II® Specimen Collection and Transport Systems
Swabs Transport Swabs
Sterile, ready to use units for the collection and transport of bacteriological specimens
Both Amies provide excellent environments for the survival of anaerobes through the complete immersion of the swabs into the gel and by the presence of the reducing agent sodium thioglycollate. Shelf life is 24 months. Amies Clear is a modified Amies gel that provides excellent survival for a wide range of bacteria and is compatible with gram stain. Amies Charcoal enhances the survival of fastidious organisms by neutralizing any toxins that may be present in the specimen.
Modified Liquid Stuart'sMore Product Information