Nunc™ Biobanking and Cell Culture Cryogenic Tubes, Thermo Scientific
Supplier: Thermo Fisher Scientific
347597 347627 351934 367997 373530 347643 337516 363401 377267 368632 366524 379189 363452 379146 366656 377224 375353 375418
CA55710-282CS 3822.49 CAD
CA55710-282 CA55710-284 CA73521-388 CA73521-390 CA73521-444 CA55710-280 CA66021-984 CA66021-985 CA66021-986A CA66021-987 CA66021-988 CA66021-989 CA66021-990A CA66021-991 CA66021-992 CA66021-993 CA66021-994 CA66021-996
Nunc™ Biobanking and Cell Culture Cryogenic Tubes, Thermo Scientific
Vials
These cryogenic storage tubes and accessories are designed to work together and provide the best solutions for the most trusted sample storage.
  • Low cellular adhesion and low protein binding
  • Non-cytotoxic, non-mutagenic plastics and pyrogen-free, IVD compatible vials ensure sample integrity
  • Internally-threaded tubes suitable for storage down to the vapor phase of liquid nitrogen
  • Externally threaded options can store at higher general refrigeration temperatures (4 °C)
Applications includes: Regulated applications, irreplaceable high value contentMore Product Information