Nunc-Immuno™ and Fluoronunc™/Luminunc™ 96-Well Modules and Frames, Thermo Scientific
Supplier: Thermo Fisher Scientific
469264 469914 469922 469949 469957 473717 475515 467466 468667 441653 469078 475086 437702 437591 437915 475523 467120 467140 473709 444865 445101 475078
CA62409-130CS 862.74 CAD
CA62409-130 CA62409-132 CA62409-133 CA62409-134 CA62409-135 CA62409-136 CA62409-137 CA62409-228 CA62409-261 CA62409-280 CA62409-283 CA62409-295 CA62409-300 CA62409-302 CA62409-306 CA62409-316 CA62409-448 CA62409-450 CA62409-287 CA62409-288 CA62409-289 CA62409-294
Nunc-Immuno™ and Fluoronunc™/Luminunc™ 96-Well Modules and Frames, Thermo Scientific
Microplates Immunology Microplates and Strips
Composed of individual modules consisting of either 8, 12, or 16 wells, these plates are designed specifically for quantitative immunoassays and solid phase assays
Well modules and frames are compatible with 96-well plate automation. Well modules are held firmly by the frame and will not come loose when inverted. Modules are made of virgin polystyrene. The mixed hydrophilic/hydrophobic MaxiSorp™ surface binds to a wide variety of biomolecules, but has been optimized to bind IgG. Recommended for assays where glycoproteins, such as antibodies, are adsorbed into the plastic. The hydrophobicMore Product Information