Nunc® MicroWell™ 96-Well Plates, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
268200 243656 262162 269787 260860
CA62407-184CS 1820.19 CAD
CA62407-184 CA62409-111 CA62409-116 CA62409-120 CA62407-174
Nunc® MicroWell™ 96-Well Plates, Sterile, Thermo Scientific
Microplates HTS Microplates
These sterile, polystyrene plates are ideal for bacteriology, serology, molecular biology, and other laboratory applications
Plates have a standard format, which makes them compatible with plate washers and readers. V-bottom and U-bottom plates allow pelleting or settling of cells for gentle concentration and sample retrieval. Flat-bottom wells can be easily examined microscopically.
The wells have raised rims to reduce risk of cross-contamination. Lids have rings to control condensation during incubation. The beveled corners on the plates and lids prevent accidental reversal of theMore Product Information