Nunc® Lab-Tek™ Chamber Slide™ Systems, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
177410 177429 177380 177399 177402 177410 177429 177437 177445 178599
62407-320CS 1974.47 CAD
62407-320 62407-325 CA62407-305 CA62407-310 CA62407-317 CA62407-320 CA62407-325 CA62407-331 CA62407-335A CA62407-350
Nunc® Lab-Tek™ Chamber Slide™ Systems, Sterile, Thermo Scientific
Slides Chamber Slides
Systems feature a removable polystyrene medium chamber so cells can be grown, fixed, and stained on the same slide.
  • Removable media wells are attached to either a soda-lime glass microscope slide or a solvent-resistant Permanox® plastic slide by a medical-grade silicone gasket
  • A convenient writing surface is easily marked, providing positive sample identification
  • Multiwell design allows researchers to analyze different specimens, immunochemicals, or stains on one slide without transfer from other vessels
Tested for quality and sterility.