Nunc™ Cell Factory™, Large Scale Production, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
164327 139446 167695 164327 170009 165250 170009 170009 140004
62407-100CS 1066.26 CAD
62407-100 CA62407-081 CA62407-099 CA62407-100 CA62407-106 CA62407-098 CA-TX62407-106 62407-106 10024-260
Nunc™ Cell Factory™, Large Scale Production, Sterile, Thermo Scientific
Bioreactors
Cell factory units maximize available growing area with low risk of contamination and easy handling
  • Polystyrene construction
  • Easy to customize and close
  • Large-scale production
Molded from high-quality polystyrene with the Nunclon™Δ tissue culture treated surface. Each cell factory is a sealed stack of chambers with common vent and fill ports. The 40-chamber factory can be used in place of 30 roller bottles. Openings connecting the chambers cause media to fill evenly for consistent growth conditions. Vents can be capped or fitted with bacterial airMore Product Information