Anti-IgL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
610-4311
CA610-4311EA 498 CAD
CA610-4311
Anti-IgL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Rabbit Anti-λ (lambda chain) Reacts with Mouse

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Light Chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol IgL
Antigen name Immunoglobulin L
Environmentally Preferable
Antigen synonyms lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
Antibody type Secondary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope Light Chain
Immunogen Mouse lambda light chain
Format Antibody fraction
Form Lyophilized
Purification purified by immunoaffinity chromatography
Concentration 1.0 mg/ml by UV absorbance at 280 nm
Cross adsorption No
Storage buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
Storage temperature –20 °C
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR