Microcentrifuge Screw Caps Closures for Micrewtubes™ Centrifuge Tubes, Simport
Supplier: Simport Scientific
T340NOS T340GOS T340LOS T340ROS T340YOS T340WOS T340BOSL T340GOSL T340LOSL T340NOSL T340ROSL T340WOSL T340YOSL T340NLS T340BOS T345AS T340BLSFT T340BRLSFT T340GLSFT T340LLSFT T340NLSFT T340OLSFT T340RLSFT T340WLSFT T340YLSFT T340BOSFT T340GOSFT T340LOSFT T340NOSFT T340OOSFT T340ROSFT T340WOSFT T340YOSFT T340BROSL T340OOSL T340BLSL T340BRLSL T340GLSL T340LLSL T340NLSL T340OLSL T340RLSL T340WLSL T340YLSL T340BLS T340BRLS T340GLS T340LLS T340OLS T340RLS T340WLS T340YLS T340BROS T340OOS T340OOS T340NLS T340BLS T340GLS T340LLS T340OLS T340RLS T340YLS T340WLS T340BRLS T340NOSL T340BOSL T340GOSL T340LOSL T340OOSL T340ROSL T340YOSL T340WOSL T340BROSL T340NLSL T340BLSL T340GLSL T340LLSL T340OLSL T340RLSL T340YLSL T340WLSL T340BRLSL T340NOSFT T340BOSFT T340GOSFT T340LOSFT T340OOSFT T340ROSFT T340YOSFT T340WOSFT T340BROSFT T340NLSFT T340BLSFT T340GLSFT T340LLSFT T340OLSFT T340RLSFT T340YLSFT T340WLSFT T340BRLSFT T345AS
CA60872-795CS 283.15 CAD
CA60872-795 CA60872-796 CA60872-797 CA60872-798 CA60872-799 CA60872-800 CA60872-820 CA60872-821 CA60872-822 CA60872-823 CA60872-824 CA60872-825 CA60872-826 CA60872-414 CA73960-272 CA11027-894 CA11028-594 CA11028-596 CA11028-598 CA11028-600 CA11028-602 CA11028-604 CA11028-606 CA11028-608 CA11028-610 CA11028-612 CA11028-616 CA11028-618 CA11028-620 CA11028-622 CA11028-624 CA11028-626 CA11028-628 CA11028-630 CA11028-632 CA11028-634 CA11028-636 CA11028-638 CA11028-640 CA11028-642 CA11028-644 CA11028-646 CA11028-648 CA11028-650 CA11028-652 CA11028-654 CA11028-656 CA11028-658 CA11028-660 CA11028-662 CA11028-664 CA11028-666 CA11028-668 CA11028-670 89499-142 89499-144 89499-146 89499-148 89499-150 89499-152 89499-154 89499-156 89499-158 89499-160 89499-162 89499-164 89499-166 89499-168 89499-170 89499-172 89499-174 89499-176 89499-178 89499-180 89499-182 89499-184 89499-186 89499-188 89499-190 89499-192 89499-194 89499-196 89499-198 89499-200 89499-202 89499-204 89499-206 89499-208 89499-210 89499-212 89499-214 89499-216 89499-218 89499-220 89499-222 89499-224 89499-226 89499-228 89499-230 89499-232 89500-344
Microcentrifuge Screw Caps Closures for Micrewtubes™ Centrifuge Tubes, Simport
Closures Test/Sample Tube Closures
Six styles of caps to choose from and two sealing types: O-Ring seal and lip seal
The silicone O-Ring caps are made of polypropylene. Will withstand temperatures from -196 to 121°C and autoclaving. The lip seal caps are made of high density polyethylene. Will withstand temperatures from -196 to 110 °C.
The cap is molded with a deep internal lip that fits snugly against the interior wall of the tube thus preventing the contents from coming in contact with the seal or threads, thereby reducing the chances of sample contamination. The cap’s high profile facilitates manipulation especiallyMore Product Information