Biotube™ Cluster Tube System, Simport
Supplier: Simport Scientific
T100-26 T100-28 T100-40 T100-2B T100-4B T100-4G T100-4R T100-20 T100-25 T100-35 T100-2G T100-2R T100-2Y T100-3B T100-3G T100-3R T100-3Y T100-4Y T100-50B T100-50G T100-50R T100-50Y T100-60B T100-60G T100-60R T100-60Y T100-1B T100-1G T100-1R T100-1Y T100-50Y T100-60B T100-60G T100-60R T100-60Y T100-50B T100-50G T100-50R T100-20LST
CA60870-021CS 323.49 CAD
CA60870-021 CA60870-026 CA60870-028 CA60872-504 CA60872-512 CA60872-513 CA60872-514 CA60872-489 CA60872-490 CA60872-494 CA60872-505 CA60872-506 CA60872-507 CA60872-508 CA60872-509 CA60872-510 CA60872-511 CA60872-515 CA60872-520 CA60872-521 CA60872-522 CA60872-523 CA60872-524 CA60872-525 CA60872-526 CA60872-527 CA60872-684 CA60872-686 CA60872-688 CA60872-690 10199-456 10199-544 10199-546 10199-548 10199-550 10199-558 10199-630 10199-632 10199-640
Biotube™ Cluster Tube System, Simport
Tubes Microtubes
The Biotube™ Cluster Tube System is a polypropylene tube and rack system with tubes held in a standard 8×12 configuration.
  • Alphanumeric coding and removeable 96-place ID card for accurate sample location
  • Transparent cover keyed to base to prevent misalignment
  • Ideal for storing, freezing and transporting reagents and specimens
  • Racks and tubes can be used over a wide temperature range, −80 to 121 °C
Tubes and racks can be autoclaved up to 100 times and will resist most research chemicals. The tube supporting grid plate can be removed and used as aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable