Falcon® Round Bottom High Clarity Polypropylene Tube, Disposable, Corning
Supplier: Corning
352053 352018 352063 352006 352059 352002
CA60819-659CS 540.64 CAD
CA60819-659 CA60819-670 CA60819-728 CA60819-740 CA60819-761 CA60819-794
Falcon® Round Bottom High Clarity Polypropylene Tube, Disposable, Corning
Tubes Culture Tubes
These disposable, clear polypropylene tubes are best suited for procedures requiring high optical clarity, and thermal and chemical stability. Provide reliable containment of laboratory fluid samples.
  • Dual position snap-cap prevents sample loss with ease of handling, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • 17×100 mm tubes have blue printed graduations and white writing patch
  • Tube dimensions and volumes are approximate
  • Widely used and referenced in laboratory protocols
  • Sterile,
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification