Anti-IgG Goat Polyclonal Antibody (Cy5.5®)

Supplier: Rockland Immunochemical
607-113-130
CA607-113-130EA 512 CAD
CA607-113-130
Anti-IgG Goat Polyclonal Antibody (Cy5.5®)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Golden Syrian Hamster
Order Now

SPECIFICATIONS

Antigen symbol IgG
Antigen name Immunoglobulin G
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Golden syrian hamster
Host Goat
Isotype
Epitope Heavy and Light chain
Concentration 1.0 mg/ml by UV absorbance at 280 nm
Storage buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR