Anti-T Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
600-403-263
CA600-403-263EA 875 CAD
CA600-403-263
Anti-T Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Phospho Site Specific Threonine affinity purified antibody is suitable for use in ELISA and western blotting assays.  The antibody reacts specifically with phosphothreonine and shows minimal reactivity by ELISA and western blotting with Proteins.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:500;

Type: Primary
Antigen: T
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR