Anti-GDF15 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-B07
CA600-401-B07EA 834 CAD
CA600-401-B07
Anti-GDF15 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody is suitable for ELISA and western blotting of human and mouse NAG-1 Protein.

Recommended Dilutions: ELISA: 1:300,000; Western Blot: 1:2000 - 1:10000;

Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR