High Torque Overhead Stirrers, 220 V
Supplier: Caframo
BDC3030220U1 BDC6015220U1 BDC1850220U1
CA58960-167EA 2995.23 CAD
CA58960-167 CA58960-173 CA58960-183
High Torque Overhead Stirrers, 220 V
Stirrers Overhead Stirrers
Designed for continuous, high viscosity mixing.
  • 220 V model
These benchtop overhead stirrers are most suitable for highly viscous mixing applications. Other applications include cosmetics and personal care products, inks and coatings, paint, pharmaceuticals, pulp and paper, pilot plants, adhesives, and epoxies.
Brushless, sparkless DC motor and two speed transmission deliver constant performance under changing conditions. Speed is maintained through electronic control as product viscosity changes. Stirrers are built with a totally enclosed metalMore Product Information