Chemical Sorbents, 3M™ Company
Supplier: 3M
7000001911 7000001947 7000001909 7000001910
CA56616-390EA 360.48 CAD
CA56616-390 CA56222-918 CA56616-386 CA56616-388
Chemical Sorbents, 3M™ Company
Spill Control Materials
For use on most aggressive (hazardous) and non-aggressive liquids.
  • Chemical folded sorbents are one of the most versatile environmental safety products available
  • Spill kit allows you to easily select the product to fit the response
  • Encircle spills and contain leaks with mini-booms
Chemical folded sorbents are one of the most versatile environmental safety products available
Spill kit combines the best of both worlds - chemical sorbent-folded (which can be used as boom, pillows or a roll) and high capacity chemical sorbent pads. This uniqueMore Product Information
Order Now
SPECIFICATIONS
Type Coolants; Water-Soluble Oils Acids/Alkalis/Alcohols; Chemicals; Flammable Liquids; Solvents - Hydrocarbon Chlorinated Water-Soluble Oils; Coolants
Environmentally Preferable