Leakbuster™ Specimen Containers
Supplier: Starplex
B952-LFS B1202-1YN B1202-VO WB902-1YN B202-1W-CO B900-1W-CO B900-1O-CO B602-VO B1202-1YNL B1202-1O-OR B1252-1O B602-1YNL B602-1YN B602L B902-1W B902-VO B902-1YN B902-1YNL V9740 B202-VO B1202-1W-CO B602-1O B1202-1O B952-1O WB902-1O B902-1O B902L B1202-VOS WB902-1WNL B1202-1W-COS
CA11006-280CS 422.39 CAD
CA11006-280 CA53594-810 CA53594-812 CA53594-824 CA55500-022 CA55500-023 CA55500-046 CA55500-053 CA68810-007 CA68810-008 CA68810-010 CA73240-092 CA73240-094 CA73240-098 CA73240-110 CA73240-116 CA73240-120 CA73240-150 CA73270-064 CA55500-031 CA55500-156 CA73240-038 CA73240-106 CA73250-004 CA73240-109L CA73240-132L CA73240-028L CA10798-884 CA10798-954 CA10799-212 CA10798-955
Leakbuster™ Specimen Containers
Sample Containers General Purpose Containers
This premium quality line of specimen containers consists of 60 ml, 90 ml, wide 90 ml, and 120 ml sizes for leakproof transportation.
Containers are ideal for use in harsh transport conditions or in pneumatic tube systems. All vials are manufactured from clear, flexible, crack-resistant polypropylene. The streamlined straight-sided vials are stable, stackable, and save space in storage. There are graduation markings on three sides. All sterile containers have a tab on the label to ensure the sterility integrity of the container. Midstream kits consist of a LeakBuster™ container and aMore Product Information