False Bottom Vials, Polypropylene, Starplex Scientific
Supplier: Starplex
V8991
CA55500-054CS 570.89 CAD
CA55500-054
False Bottom Vials, Polypropylene, Starplex Scientific
Vials
Vials are made of crack-resistant medical grade polypropylene.
Vials can be used in many different types of automated analyzer systems. The label placement line, which is located just below the threads, allows accurate placement of a bar code label prior to the vial being capped. The opaque band around the bottom of the vial provides a reference for orientation by an automated system. The vial bottom is conical to prevent probe crashing, and tapered to ensure easy placement into racking.
Screw caps should be used when transporting the vial.
Available clear for easy visual inspectionMore Product Information