Vacushield™ Vent Device, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
4402
CA55095-006PK 112.18 CAD
CA55095-006
Vacushield™ Vent Device, Cytiva (Formerly Pall Lab)
Filters Venting Filters
This disposable protective device is designed to prevent aqueous liquid and aerosol contamination of vacuum pumps or aspiration suction systems.
  • Disposable
  • High air flow
  • High bacterial retention
It provides high air flow rates and bacterial retention without loss of pump efficiency. Non-sterile. May be autoclaved or ETO sterilized. Hydrophobic, microporous PTFE membrane filter achieves 99.97% D.O.P. retention (0.3 µm particles in air). Features polypropylene housing and 6.4 - 12.7 mm (¹/₄ - ¹/₂") stepped hose barb connections (internal taper acceptsMore Product Information