Centrifuge Tubes, 50 ml, Simport Scientific
Supplier: Simport Scientific
T420-4 T420-7 T420-1 T420-3 T420-5 T420-7 T420-1 T420-3 T420-5
CA60829-020CS 490.49 CAD
CA60829-020 CA60829-032 CA53594-083 CA53594-084 CA53594-086 490010-464 76299-294 76299-296 76299-298
Centrifuge Tubes, 50 ml, Simport Scientific
Tubes Centrifuge Tubes
These centrifuge tubes are useful for collecting and transporting biological specimens.
  • Polypropylene or polystyrene tube options
  • Sterile or non sterile versions available
  • Cap features a leakproof design
  • Polypropylene tubes are autoclavable
Leakproof characteristics are ensured by a flat top plastic screw cap with an inner sealing lip. Tubes are made of translucent polypropylene or optically clear polystyrene with molded graduations from 2.5 to 50 ml. Polypropylene tubes can be autoclaved and will resist temperatures up to 121 °C and they will alsoMore Product Information