Eppendorf® 5804 Benchtop Centrifuge
Supplier: Eppendorf
022622501 022628135 022628034 022628203 022628149 022628048 022628214
CA53513-800EA 11767.26 CAD
CA53513-800 CA53513-802 CA89210-968 CA89210-970 CA89210-972 89305-158 CA11017-350 CA11017-354 490013-046
Eppendorf® 5804 Benchtop Centrifuge
Centrifuges Benchtop Centrifuges
The high-speed Eppendorf Centrifuge 5804 is a medium capacity centrifuge, ideal for molecular applications in tubes up to 250 ml. It also offers supreme versatility with the capability of taking swing-bucket and fixed-angle rotors, as well as deepwell plates.
  • Low access height of 29 cm for easy loading and unloading of samples
  • Quiet operation to improve work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • Swing-bucket rotors and adapters accommodate tubes and
More Product Information