CrossLab Ultra Inert Liners for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5190-2293 5190-2294 5190-2295 5190-3163 5190-3164 5190-3165 5190-3167 5190-3169 5190-3171 5190-3983 5190-4007 5190-4047 5190-4048 5190-6168 5190-2292 5190-3162 5190-3166 5190-3168 5190-3172 5190-4056 5190-5112 5190-7020 5190-5112-005 5190-3170 5190-3173 5190-5105 5190-5105-005 5190-7012 5190-7013 5190-7014 5190-7045
CA5190-2293EA 82.32 CAD
CA5190-2293 CA5190-2294 CA5190-2295 CA5190-3163 CA5190-3164 CA5190-3165 CA5190-3167 CA5190-3169 CA5190-3171 CA5190-3983 CA5190-4007 CA5190-4047 CA5190-4048 CA5190-6168 CA5190-2292 CA5190-3162 CA5190-3166 CA5190-3168 CA5190-3172 CA5190-4056 CA5190-5112 CA5190-7020 CA51905112005 CAAG5190-3170 CAAG5190-3173 CAAG5190-5105 CA51905105005 CAAG5190-7012 CAAG5190-7013 CAAG5190-7014 CAAG5190-7045
CrossLab Ultra Inert Liners for GC, Agilent Technologies
Chromatography Supplies Chromatography Inlet Liners
Agilent Ultra Inert inlet liners prevent adsorption and ensures accurate sample transfer onto the GC column.
  • Glass-fritted liners now available for splitless and universal applications to help increase reproducibility and extend liner life
  • Consistent high level of inertness for greater sensitivity, accuracy, and reproducibility
  • Ultra Inert split and splitless liners, with and without wool, for a wide variety of applications
  • Liners with inert wool retain nonvolatiles, making trace-level analysis of difficult active compounds easier
  • Unique touchless
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable