Liner O-Rings for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0905-1145 5061-5867 5180-4173 5180-4181 5080-4982 5180-4168 5188-5365 5190-2269 5190-2268 MKI-U-COV10 0905-1608 0905-1609 0905-1610 5188-5311 392611940
CA0905-1145PK 46.36 CAD
CA0905-1145 CA5061-5867 CA5180-4173 CA5180-4181 CA5080-4982 CA5180-4168 CA76508-184 CA5190-2269 CA5190-2268 CAMKI-U-COV10 CAAG0905-1608 CAAG0905-1609 CAAG0905-1610 CAAG5188-5311 AG392611940
Liner O-Rings for GC
Seals Sealing Rings
Liner O-rings for GC and GC/MS systems.
  • Liners are sealed in the inlet with O-rings or graphite seals
  • Graphite seals are used when inlet temperatures exceed 350 °C
  • Fluorocarbon O-ring seals are easier to replace than graphite that deforms and flakes apart
  • Plasma treated for a nonstick, contaminant-free surface that won’t stick to the inlet metal surface
  • Packaged for convenience and cleanliness in a novel dial package that delivers 1 clean O-ring at a time.
Liner O-rings are used to form a proper seal around the outside of the inlet liner between theMore Product Information