CrossLab Programmable Temperature Vaporizing (PTV) Inlet for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5182-9747 5182-9748 5182-9759 5182-9761 5182-9763 5182-9768 5182-9769 5183-0318 5183-2036 5183-2038 5183-2058 G1544-80550 18740-60835 5182-9749 5183-2037 9301-0713 9301-0892 3150-0602 5062-3525 5182-9751 5182-9754 5182-9756 5182-9757 5182-9758 5182-9760 5182-9762 5182-9770 5182-9775 5188-5313 5188-5356 5190-1426 G2617-60507 5021-7133 5021-7134 5021-7136 5021-7137
CA5182-9747EA 565.09 CAD
CA5182-9747 CA5182-9748 CA5182-9759 CA5182-9761 CA5182-9763 CA5182-9768 CA5182-9769 CA5183-0318 CA5183-2036 CA5183-2038 CA5183-2058 CAG1544-80550 CA76514-568 CA76514-812 CA76514-816 CA76515-128 CA76515-136 CAAG3150-0602 CAAG5062-3525 CAAG5182-9751 CAAG5182-9754 CAAG5182-9756 CAAG5182-9757 CAAG5182-9758 CAAG5182-9760 CAAG5182-9762 CAAG5182-9770 CAAG5182-9775 CAAG5188-5313 CAAG5188-5356 CAAG5190-1426 CAAGG261760507 AG5021-7133 AG5021-7134 AG5021-7136 AG5021-7137
CrossLab Programmable Temperature Vaporizing (PTV) Inlet for GC
Chromatography Supplies Chromatography Spare Parts
Agilent programmed temperature vaporizer inlets combine the benefits of split, splitless, and on-column inlets, resulting in less thermal degradation of labile compounds.
  • Perform large volume injections for low minimum detection limits
  • Get less thermal degradation of labile compounds
Agilent programmed temperature vaporizer inlets combine the benefits of split, splitless, and on-column inlets. The sample is usually injected into a cool liner, so syringe needle discrimination does not occur. Then the inlet temperature is increased to vaporize theMore Product Information