Anti-G-CSF Rat Monoclonal Antibody (Biotin) [clone: BVD11-37G10]

Supplier: Biolegend
506702
CA506702-BLEA 523.42 CAD
CA506702-BL
Anti-G-CSF Rat Monoclonal Antibody (Biotin) [clone: BVD11-37G10]
Antibodies
Biotin anti-human G-CSF [BVD11-37G10]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 1-4 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: G-CSF
Clonality: Monoclonal
Clone: BVD11-37G10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR