SlideFolder™ Microscope Slide Holders, Simport Scientific
Supplier: Simport Scientific
M750-20B M750-20G M750-20W M750-20Y M750-20GY M750-20AS M752-20WOP
CA48648-530CS 255.73 CAD
CA48648-530 CA48648-532 CA48648-536 CA48648-538 CA48648-534 CA48648-540 CA89039-698
SlideFolder™ Microscope Slide Holders, Simport Scientific
Slide Holders
These two-part folders hold up to twenty standard microscope slides
  • All units are stackable
  • Not autoclavable
The high-impact polystyrene base holds slides horizontally with numbered spaces for easy identification. The transparent polystyrene door covers the slides and can be swung behind the folder for space-saving purposes.
Temperature range: –80 to 80°C (–112 to 176°F)