VWR® Micro Cover Glasses, Round, No. 1
Supplier: VWR International
48380-080 CA48380-046 CA48380-068
48380-080CS 1644.07 CAD
48380-080 CA48380-046 CA48380-068
VWR® Micro Cover Glasses, Round, No. 1
Cover Glasses
These cover glasses offer maximum clarity, superior corrosion resistance, and uniform thickness and size.
  • Circular. No. 1 micro cover glasses have a thickness of 0.13 to 0.17mm
They are free of striae and oily film.
Ordering information: Packaged 1 oz. per plastic box.
Order Now
SPECIFICATIONS
Environmentally Preferable