Formaldehyde 37% aqueous solution Stabilised ACS

Supplier: RICHARD-ALLAN SCIENTIFIC CA
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilization: MeOH

9300-5 9300-1
CA48218-722EA 528.91 CAD
CA48218-722 CA48218-724
Formaldehyde 37% aqueous solution Stabilised ACS
Formaldehyde

Not intended for use as a disinfectant.

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 2209
ADR: 8 III
Merck Index: 13,04259

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR