Rotors and Accessories for Eppendorf® Compact Centrifuges
Supplier: Eppendorf
022639277 022639528 022639447 022639439 022639382 022639391 022639145 022639161 022639293 022639081 022639200 022639129 022639188 022639226 022639285 022639102 022639510 022639455 022639307 022639323 022639340 022639366 022639242 022639269 022639188 022639285 5702763004
CA022639277PK 775.9 CAD
CA022639277 CA47743-478 CA47743-484 CA47743-486 CA47743-498 CA47743-500 CA47743-502 CA47743-504 CA47743-510 CA47743-512 CA47743-514 CA47743-516 CA47743-518 CA47743-520 CA47743-524 CA47743-526 CA47743-476 CA47743-488 CA47743-490 CA47743-492 CA47743-494 CA47743-496 CA47743-506 CA47743-522 47743-518 47743-524 CA11017-486
Rotors and Accessories for Eppendorf® Compact Centrifuges
Centrifuges Rotors
For use with 47743-472 series centrifuges
Rotors may be used with all centrifuge models in the series. Rotors and accessories are autoclavable to 121 °C (250 °F) for 20 minutes.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification Renewable Energy Certificate, ISO 14001