PROSAT® Polynit Heatseal Wipes, Contec®
Supplier: Contec
PSPS0070
CA47732-732CS 1555.98 CAD
CA47732-732
PROSAT® Polynit Heatseal Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
PROSAT® Polynit Heatseal Wipes are manufactured from 100% knitted textured polyester with laser-cut sealed edges for very low levels of particles and fibers. Polyester wipes are chemically resistant and exceptionally low in particles and fibers. An interlock knit creates a durable fabric, and the addition of a periodic additional stitch of a no-run interlock knit prevents the fabric from unraveling. This creates an even more durable fabric with strong edges.
  • Recommended for ISO Class 3–5 environments
  • Presaturated with 90% semi conductor grade isopropanol
More Product Information