pH Electrode, PEI Body Field Application
Supplier: Hanna
HI12300
CA470349-830EA 201.43 CAD
CA470349-830
pH Electrode, PEI Body Field Application
Electrodes pH Electrodes
This pH electrode is a plastic body, double junction, gel filled pH electrode with a built-in temperature sensor for temperature compensated measurements in a single probe design.
  • Spherical glass tip
  • Double junction reference
  • 3.5 mm connector
This probe features a 3.5 mm connector designed exclusively to be used with Hanna Instruments edge® Tablet Meters. It also features a single ceramic junction, and the spherical pH sensing portion is made with general purpose glass. The electrode has a rugged PEI (polyetherimide) body that has the chemicalMore Product Information