Fly Life Cycle Plastomount
Supplier: ED SPELDY EAST CO. LTD. CA
TL05
CA470324-664EA 99.6 CAD
CA470324-664
Fly Life Cycle Plastomount
Specimens Preserved Specimens Preserved Animal Materials
Preserved specimens in resin material.
  • High quality display mount
This kit looks at the Life Cycle of the Fly. The Fly undergoes complete metamorphosis. There is one specimen block included. Specimens are preserved and encased in clear resin material. They can be observed from every side to see details.
Specimens include the different stages of the Onion Fly (Delia antiqua).
Delivery information: Teacher information guide, fact sheet, student activity sheet, and answer key are included.