Rabbit Dissection Plastomount
Supplier: ED SPELDY EAST CO. LTD. CA
T608
CA470324-576EA 496.7 CAD
CA470324-576
Rabbit Dissection Plastomount
Specimens Preserved Specimens Preserved Animal Materials
Preserved specimens in resin material.
  • High quality display mount
This block looks at a real rabbit dissection. There is one specimen block included. The specimen is preserved and encased in clear resin material. It can be observed from every side to see details.
The specimen is a rabbit (Lepus sinensis).
Delivery information: Teacher information guide, student activity sheet and answer key are included.