Amino Labs BioInk Bacterial Paint Kits
Supplier: AMINO LABS NORTH INCORPORATED CA SE
WWA011-G_CA WWA011-S_CA
CA470318-708EA 827.59 CAD
CA470318-708 CA470318-710
Amino Labs BioInk Bacterial Paint Kits
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Create prints and textile art with colorful bacteria.
  • Safety First! The kit uses a non-pathogenic strain of E. coli (K-12)
  • Recommended for use with Amino Labs DNA Playground™ (available separately)
Grow patterns, paintings and symbols on petri dishes using the colored bacteria provided in your kit (as well as any colored engineered bacteria you may have created before). Once grown, transfer them directly to paper or textile in any design you like, or use some of the paintbrushes to add some freehand elements to your art. Finish by stabilizing andMore Product Information